RIE156326441 Rieke Metals, Inc.

cyclopentyl 2-(thiomorpholinomethyl)phenyl ketone

  • MDL number MFCD03842793
  • Cas Number : 898782-56-4
  • Molecular Weight : 289.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0703-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0703-00
Availability details for
Quantity
  • Availability
  • Add to Cart