RIE156326442 Rieke Metals, Inc.

cyclohexyl 2-(thiomorpholinomethyl)phenyl ketone

  • MDL number MFCD03842794
  • Cas Number : 898782-58-6
  • Molecular Weight : 303.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0704-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0704-00
Availability details for
Quantity
  • Availability
  • Add to Cart