RIE156326445 Rieke Metals, Inc.

ethyl 6-oxo-6-[2-(thiomorpholinomethyl)phenyl]hexanoate

  • MDL number MFCD03842797
  • Cas Number : 898782-64-4
  • Molecular Weight : 349.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0707-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0707-00
Availability details for
Quantity
  • Availability
  • Add to Cart