RIE156326448 Rieke Metals, Inc.

2-(4-methylpiperazinomethyl)benzophenone

  • MDL number MFCD03842800
  • Cas Number : 24088-65-1
  • Molecular Weight : 294.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-01
Availability details for
Quantity
  • Availability
  • Add to Cart