RIE156326450 Rieke Metals, Inc.

3'-methyl-2-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842802
  • Cas Number : 898782-72-4
  • Molecular Weight : 308.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-03
Availability details for
Quantity
  • Availability
  • Add to Cart