RIE156326453 Rieke Metals, Inc.

3'-methoxy-2-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842805
  • Cas Number : 898782-78-0
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl){2-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C20H24N2O2/c1-21-10-12-22(13-11-21)15-17-6-3-4-9-19(17)20(23)16-7-5-8-18(14-16)24-2/h3-9,14H,10-13,15H2,1-2H3
Inchi Key WJIASZDEUZRVJD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0800-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-06
Availability details for
Quantity
  • Availability
  • Add to Cart