RIE156326454 Rieke Metals, Inc.

4'-methoxy-2-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842806
  • Cas Number : 898782-80-4
  • Molecular Weight : 324.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-07
Availability details for
Quantity
  • Availability
  • Add to Cart