RIE156326461 Rieke Metals, Inc.

2-(4-methylpiperazinomethyl)-2'-thiomethylbenzophenone

  • MDL number MFCD03842813
  • Cas Number : 898782-97-3
  • Molecular Weight : 340.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-14
Availability details for
Quantity
  • Availability
  • Add to Cart