RIE156326463 Rieke Metals, Inc.

3'-bromo-2-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842815
  • Cas Number : 898783-01-2
  • Molecular Weight : 373.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-18
Availability details for
Quantity
  • Availability
  • Add to Cart