RIE156326468 Rieke Metals, Inc.

4'-fluoro-2-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842820
  • Cas Number : 898783-11-4
  • Molecular Weight : 312.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-23
Availability details for
Quantity
  • Availability
  • Add to Cart