RIE156326470 Rieke Metals, Inc.

2,4-dimethyl-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842822
  • Cas Number : 898783-15-8
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-27
Availability details for
Quantity
  • Availability
  • Add to Cart