RIE156326471 Rieke Metals, Inc.

2,5-dimethyl-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842823
  • Cas Number : 898783-17-0
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-28
Availability details for
Quantity
  • Availability
  • Add to Cart