RIE156326473 Rieke Metals, Inc.

3,4-dimethyl-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842825
  • Cas Number : 898783-21-6
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-30
Availability details for
Quantity
  • Availability
  • Add to Cart