RIE156326475 Rieke Metals, Inc.

4-bromo-3-fluoro-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842827
  • Cas Number : 898783-25-0
  • Molecular Weight : 391.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-32
Availability details for
Quantity
  • Availability
  • Add to Cart