RIE156326476 Rieke Metals, Inc.

4-chloro-3-fluoro-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842828
  • Cas Number : 898783-27-2
  • Molecular Weight : 346.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-33
Availability details for
Quantity
  • Availability
  • Add to Cart