RIE156326478 Rieke Metals, Inc.

2-chloro-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842830
  • Cas Number : 898783-31-8
  • Molecular Weight : 328.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-36
Availability details for
Quantity
  • Availability
  • Add to Cart