RIE156326480 Rieke Metals, Inc.

2-(4-methylpiperazinomethyl)-2'-trifluoromethylbenzophenone

  • MDL number MFCD03842832
  • Cas Number : 898783-35-2
  • Molecular Weight : 362.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {2-[(4-methyl-1-piperazinyl)methyl]phenyl}[2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C20H21F3N2O/c1-24-10-12-25(13-11-24)14-15-6-2-3-7-16(15)19(26)17-8-4-5-9-18(17)20(21,22)23/h2-9H,10-14H2,1H3
Inchi Key LBCHWGCIMIQCMU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0800-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-43
Availability details for
Quantity
  • Availability
  • Add to Cart