RIE156326481 Rieke Metals, Inc.

2-(4-methylpiperazinomethyl)-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842833
  • Cas Number : 898783-37-4
  • Molecular Weight : 362.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-44
Availability details for
Quantity
  • Availability
  • Add to Cart