RIE156326484 Rieke Metals, Inc.

2-chloro-4-fluoro-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842836
  • Cas Number : 898762-21-5
  • Molecular Weight : 346.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-50
Availability details for
Quantity
  • Availability
  • Add to Cart