RIE156326486 Rieke Metals, Inc.

4-chloro-2-fluoro-2'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842838
  • Cas Number : 898762-27-1
  • Molecular Weight : 346.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0800-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0800-52
Availability details for
Quantity
  • Availability
  • Add to Cart