RIE156326497 Rieke Metals, Inc.

cyclobutyl 2-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03842849
  • Cas Number : 898762-60-2
  • Molecular Weight : 272.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0802-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0802-00
Availability details for
Quantity
  • Availability
  • Add to Cart