RIE156326499 Rieke Metals, Inc.

cyclohexyl 2-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03842851
  • Cas Number : 898762-65-7
  • Molecular Weight : 300.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0804-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0804-00
Availability details for
Quantity
  • Availability
  • Add to Cart