RIE156326500 Rieke Metals, Inc.

ethyl 4-[2-(4-methylpiperazinomethyl)phenyl]-4-oxobutyrate

  • MDL number MFCD03842852
  • Cas Number : 898762-67-9
  • Molecular Weight : 318.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0805-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0805-00
Availability details for
Quantity
  • Availability
  • Add to Cart