RIE156326501 Rieke Metals, Inc.

ethyl 5-[2-(4-methylpiperazinomethyl)phenyl]-5-oxovalerate

  • MDL number MFCD03842853
  • Cas Number : 898762-69-1
  • Molecular Weight : 332.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0806-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0806-00
Availability details for
Quantity
  • Availability
  • Add to Cart