RIE156326508 Rieke Metals, Inc.

4'-methyl-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842860
  • Cas Number : 898762-83-9
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl](4-methylphenyl)methanone
Inchi Code 1S/C19H19NO/c1-15-8-10-16(11-9-15)19(21)18-7-3-2-6-17(18)14-20-12-4-5-13-20/h2-11H,12-14H2,1H3
Inchi Key VXTHILSCADLDCL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0900-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-04
Availability details for
Quantity
  • Availability
  • Add to Cart