RIE156326511 Rieke Metals, Inc.

4'-methoxy-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842863
  • Cas Number : 898762-89-5
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-07
Availability details for
Quantity
  • Availability
  • Add to Cart