RIE156326513 Rieke Metals, Inc.

3'-cyano-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842865
  • Cas Number : 898762-93-1
  • Molecular Weight : 288.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-09
Availability details for
Quantity
  • Availability
  • Add to Cart