RIE156326514 Rieke Metals, Inc.

4'-cyano-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842866
  • Cas Number : 898762-95-3
  • Molecular Weight : 288.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-10
Availability details for
Quantity
  • Availability
  • Add to Cart