RIE156326517 Rieke Metals, Inc.

4'-carboethoxy-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842869
  • Cas Number : 898762-99-7
  • Molecular Weight : 335.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-13
Availability details for
Quantity
  • Availability
  • Add to Cart