RIE156326519 Rieke Metals, Inc.

2-(3-pyrrolinomethyl)-4'-thiomethylbenzophenone

  • MDL number MFCD03842871
  • Cas Number : 898763-04-7
  • Molecular Weight : 309.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-15
Availability details for
Quantity
  • Availability
  • Add to Cart