RIE156326520 Rieke Metals, Inc.

3'-bromo-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842872
  • Cas Number : 898763-06-9
  • Molecular Weight : 342.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)[2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C18H16BrNO/c19-16-8-5-7-14(12-16)18(21)17-9-2-1-6-15(17)13-20-10-3-4-11-20/h1-9,12H,10-11,13H2
Inchi Key SEOBFOHKAQYQKQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0900-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-18
Availability details for
Quantity
  • Availability
  • Add to Cart