RIE156326525 Rieke Metals, Inc.

4'-fluoro-2-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842877
  • Cas Number : 898763-17-2
  • Molecular Weight : 281.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-23
Availability details for
Quantity
  • Availability
  • Add to Cart