RIE156326526 Rieke Metals, Inc.

2,3-dimethyl-2'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842878
  • Cas Number : 898763-20-7
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl](2,3-dimethylphenyl)methanone
Inchi Code 1S/C20H21NO/c1-15-8-7-11-18(16(15)2)20(22)19-10-4-3-9-17(19)14-21-12-5-6-13-21/h3-11H,12-14H2,1-2H3
Inchi Key LERGEKKFGJEQIV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0900-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-26
Availability details for
Quantity
  • Availability
  • Add to Cart