RIE156326529 Rieke Metals, Inc.

2,6-dimethyl-2'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842881
  • Cas Number : 898763-29-6
  • Molecular Weight : 291.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-29
Availability details for
Quantity
  • Availability
  • Add to Cart