RIE156326532 Rieke Metals, Inc.

4-bromo-3-fluoro-2'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842884
  • Cas Number : 898763-38-7
  • Molecular Weight : 360.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-32
Availability details for
Quantity
  • Availability
  • Add to Cart