RIE156326535 Rieke Metals, Inc.

2-chloro-2'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842887
  • Cas Number : 898763-47-8
  • Molecular Weight : 297.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)[2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C18H16ClNO/c19-17-10-4-3-9-16(17)18(21)15-8-2-1-7-14(15)13-20-11-5-6-12-20/h1-10H,11-13H2
Inchi Key VXOSCJREOHZSLH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0900-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-36
Availability details for
Quantity
  • Availability
  • Add to Cart