RIE156326538 Rieke Metals, Inc.

2-(3-pyrrolinomethyl)-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842890
  • Cas Number : 898763-56-9
  • Molecular Weight : 331.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-44
Availability details for
Quantity
  • Availability
  • Add to Cart