RIE156326542 Rieke Metals, Inc.

3-chloro-5-fluoro-2'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842894
  • Cas Number : 898763-66-1
  • Molecular Weight : 315.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0900-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0900-51
Availability details for
Quantity
  • Availability
  • Add to Cart