RIE156326553 Rieke Metals, Inc.

cyclopropyl 2-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842905
  • Cas Number : 898763-88-7
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl[2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]methanone
Inchi Code 1S/C15H17NO/c17-15(12-7-8-12)14-6-2-1-5-13(14)11-16-9-3-4-10-16/h1-6,12H,7-11H2
Inchi Key FQRUBCMDSSYWIZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0901-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0901-00
Availability details for
Quantity
  • Availability
  • Add to Cart