RIE156326555 Rieke Metals, Inc.

cyclopentyl 2-(3-pyrrolinomethyl)phenyl ketone

  • MDL number MFCD03842907
  • Cas Number : 898763-92-3
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0903-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0903-00
Availability details for
Quantity
  • Availability
  • Add to Cart