RIE156326558 Rieke Metals, Inc.

ethyl 5-oxo-5-[2-(3-pyrrolinomethyl)phenyl]valerate

  • MDL number MFCD03842910
  • Cas Number : 898763-98-9
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-[2-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl]-5-oxopentanoate
Inchi Code 1S/C18H23NO3/c1-2-22-18(21)11-7-10-17(20)16-9-4-3-8-15(16)14-19-12-5-6-13-19/h3-6,8-9H,2,7,10-14H2,1H3
Inchi Key HBGOYNBBWBSEIY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0906-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0906-00
Availability details for
Quantity
  • Availability
  • Add to Cart