RIE156326561 Rieke Metals, Inc.

ethyl 8-oxo-8-[2-(3-pyrrolinomethyl)phenyl]octanoate

  • MDL number MFCD03842913
  • Cas Number : 898764-04-0
  • Molecular Weight : 343.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0909-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0909-00
Availability details for
Quantity
  • Availability
  • Add to Cart