RIE156326568 Rieke Metals, Inc.

4'-carboethoxy-3-phenylpropiophenone

  • MDL number MFCD03842923
  • Cas Number : 898764-15-3
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-13
Availability details for
Quantity
  • Availability
  • Add to Cart