RIE156326570 Rieke Metals, Inc.

3'-bromo-3-phenylpropiophenone

  • MDL number MFCD03842926
  • Cas Number : 866821-66-1
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-18
Availability details for
Quantity
  • Availability
  • Add to Cart