RIE156326571 Rieke Metals, Inc.

3'-chloro-3-phenylpropiophenone

  • MDL number MFCD03842927
  • Cas Number : 388091-63-2
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-20
Availability details for
Quantity
  • Availability
  • Add to Cart