RIE156326572 Rieke Metals, Inc.

3'-fluoro-3-phenylpropiophenone

  • MDL number MFCD03842929
  • Cas Number : 898764-21-1
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-22
Availability details for
Quantity
  • Availability
  • Add to Cart