RIE156326574 Rieke Metals, Inc.

2',4'-dimethyl-3-phenylpropiophenone

  • MDL number MFCD03842932
  • Cas Number : 898764-27-7
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-27
Availability details for
Quantity
  • Availability
  • Add to Cart