RIE156326582 Rieke Metals, Inc.

2'-fluoro-3-phenylpropiophenone

  • MDL number MFCD03842941
  • Cas Number : 898764-48-2
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-37
Availability details for
Quantity
  • Availability
  • Add to Cart