RIE156326583 Rieke Metals, Inc.

3-phenyl-2'-trifluoromethylpropiophenone

  • MDL number MFCD03842942
  • Cas Number : 898764-51-7
  • Molecular Weight : 278.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-43
Availability details for
Quantity
  • Availability
  • Add to Cart