RIE156326584 Rieke Metals, Inc.

3-phenyl-3'-trifluoromethylpropiophenone

  • MDL number MFCD03842943
  • Cas Number : 67082-01-3
  • Molecular Weight : 278.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-44
Availability details for
Quantity
  • Availability
  • Add to Cart